siRNA Details
Gene Symbol: ANAPC11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002244.2 | NP_001002244.1 | 4 | 976 | ||
NM_001002245.2 | NP_001002245.1 | 3 | 645 | |||
NM_001002246.2 | NP_001002246.1 | 4 | 708 | |||
NM_001002247.2 | NP_001002247.1 | 3 | 653 | |||
NM_001002248.2 | NP_001002248.1 | 4 | 716 | |||
NM_001002249.2 | NP_001002249.1 | 4 | 767 | |||
NM_001289414.1 | NP_001276343.1 | 4 | 649 | |||
NM_001289415.1 | NP_001276344.1 | 3 | 675 | |||
NM_001289416.1 | NP_001276345.1 | 3 | 704 | |||
NM_001289417.1 | NP_001276346.1 | 5 | 800 | |||
NM_001289418.1 | NP_001276347.1 | 3 | 683 | |||
NM_001289419.1 | NP_001276348.1 | 4 | 754 | |||
NM_001289420.1 | NP_001276349.1 | 5 | 853 | |||
NM_016476.11 | NP_057560.8 | 4 | 738 | |||
GenBank mRNA | AK308091.1 | - | 2 | 559 | ||
BC171892.1 | - | 4 | 650 | |||
BC171898.1 | - | 3 | 617 | |||
BC171899.1 | - | 4 | 658 | |||
BC171900.1 | - | 3 | 587 | |||
BU554167.1 | - | 4 | 718 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
