siRNA Details
Gene Symbol: SIRT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017524.2 | NP_001017524.1 | 2 | 224 | ||
NM_012239.5 | NP_036371.1 | 2 | 370 | |||
XM_005252835.1 | XP_005252892.1 | 2 | 370 | |||
XM_011519956.1 | XP_011518258.1 | 2 | 1162 | |||
XM_011519957.1 | XP_011518259.1 | 2 | 991 | |||
XM_017017428.1 | XP_016872917.1 | 2 | 216 | |||
XM_017017429.1 | XP_016872918.1 | 2 | 390 | |||
XM_017017430.1 | XP_016872919.1 | 2 | 384 | |||
XM_017017431.1 | XP_016872920.1 | 2 | 115 | |||
GenBank mRNA | AF083108.2 | - | 2 | 438 | ||
AK299009.1 | - | 2 | 360 | |||
AK299438.1 | - | 2 | 202 | |||
AK301979.1 | - | 2 | 358 | |||
AK308681.1 | - | 2 | 371 | |||
AK316175.1 | - | 2 | 274 | |||
AL535769.3 | - | 2 | 205 | |||
BC001042.2 | - | 2 | 353 | 2823471 | ||
BI755839.1 | - | 2 | 170 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
