siRNA Details
Gene Symbol: SIRT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017524.2 | NP_001017524.1 | 6, 7 | 1060 | ||
NM_012239.5 | NP_036371.1 | 6, 7 | 1206 | |||
XM_017017430.1 | XP_016872919.1 | 6, 7 | 1220 | |||
GenBank mRNA | AF083108.2 | - | 6, 7 | 1274 | ||
AK298572.1 | - | 6, 7 | 955 | |||
AK299009.1 | - | 5, 6 | 1034 | |||
AK299438.1 | - | 6, 7 | 1038 | |||
AK316175.1 | - | 6, 7 | 1110 | |||
AL137276.1 | - | 2, 3 | 329 | |||
BC001042.2 | - | 6, 7 | 1189 | 2823471 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
