siRNA Details
Gene Symbol: C1orf56 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017860.3 | NP_060330.2 | 1 | 450 | ||
GenBank mRNA | AK000526.1 | - | 1 | 459 | ||
AK301712.1 | - | 1 | 522 | |||
AK315680.1 | - | 1 | 462 | |||
AY358436.1 | - | 1 | 493 | |||
BC002469.2 | - | 1 | 450 | 3347975 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
