siRNA Details
Gene Symbol: EML6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039753.2 | NP_001034842.2 | 36 | 5354 | ||
XM_017004098.1 | XP_016859587.1 | 38 | 5531 | |||
XM_017004099.1 | XP_016859588.1 | 37 | 5516 | |||
XM_017004100.1 | XP_016859589.1 | 38 | 5533 | |||
XM_017004101.1 | XP_016859590.1 | 32 | 4365 | |||
GenBank mRNA | AK124553.1 | - | 13 | 1384 | ||
AK131467.1 | - | 6 | 941 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_146016, AK081418, BC024726.
