siRNA Details
Gene Symbol: EML6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039753.2 | NP_001034842.2 | 32, 33 | 4755 | ||
XM_017004098.1 | XP_016859587.1 | 34, 35 | 4932 | |||
XM_017004099.1 | XP_016859588.1 | 33, 34 | 4917 | |||
XM_017004100.1 | XP_016859589.1 | 34, 35 | 4934 | |||
XM_017004101.1 | XP_016859590.1 | 28, 29 | 3766 | |||
GenBank mRNA | AK124553.1 | - | 9, 10 | 882 | ||
AK131467.1 | - | 2, 3 | 342 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
