siRNA Details
Gene Symbol: PKP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005476.2 | NP_001005476.1 | 5 | 746 | ||
NM_001304969.1 | NP_001291898.1 | 5 | 746 | |||
NM_001304970.1 | NP_001291899.1 | 5 | 746 | |||
NM_001304971.1 | NP_001291900.1 | 5 | 746 | |||
NM_003628.4 | NP_003619.2 | 5 | 746 | |||
XM_011512016.1 | XP_011510318.1 | 5 | 1183 | |||
XM_011512020.2 | XP_011510322.1 | 5 | 530 | |||
XM_011512021.2 | XP_011510323.1 | 5 | 530 | |||
XM_011512022.1 | XP_011510324.1 | 4 | 698 | |||
XM_011512026.2 | XP_011510328.1 | 5 | 530 | |||
XM_017005122.1 | XP_016860611.1 | 5 | 1183 | |||
XM_017005123.1 | XP_016860612.1 | 5 | 1718 | |||
XM_017005125.1 | XP_016860614.1 | 5 | 530 | |||
XM_017005127.1 | XP_016860616.1 | 5 | 1183 | |||
XM_017005128.1 | XP_016860617.1 | 5 | 530 | |||
XM_017005130.1 | XP_016860619.1 | 4 | 698 | |||
XM_017005132.1 | XP_016860621.1 | 3 | 270 | |||
XM_017005133.1 | XP_016860622.1 | 5 | 700 | |||
XM_017005134.1 | XP_016860623.1 | 4 | 1070 | |||
XM_017005135.1 | XP_016860624.1 | 3 | 270 | |||
GenBank mRNA | AK054911.1 | - | 5 | 1260 | ||
BC048013.1 | - | 5 | 662 | |||
BC050308.1 | - | 5 | 528 | 5262792 | ||
X81889.1 | - | 5 | 543 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
