siRNA Details
Gene Symbol: UNC80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032504.1 | NP_115893.1 | 39 | 6077 | ||
NM_182587.3 | NP_872393.3 | 39 | 6062 | |||
XM_005246476.1 | XP_005246533.1 | 40 | 6272 | |||
XM_011511004.1 | XP_011509306.1 | 40 | 6317 | |||
XM_011511005.1 | XP_011509307.1 | 40 | 6314 | |||
XM_011511006.1 | XP_011509308.1 | 40 | 6311 | |||
XM_011511007.1 | XP_011509309.1 | 40 | 6281 | |||
XM_011511008.1 | XP_011509310.1 | 40 | 6317 | |||
XM_011511010.2 | XP_011509312.1 | 40 | 6317 | |||
XM_017003884.1 | XP_016859373.1 | 40 | 6302 | |||
XM_017003885.1 | XP_016859374.1 | 40 | 6275 | |||
XM_017003886.1 | XP_016859375.1 | 40 | 6317 | |||
XM_017003887.1 | XP_016859376.1 | 39 | 6113 | |||
XM_017003888.1 | XP_016859377.1 | 39 | 6071 | |||
XM_017003889.1 | XP_016859378.1 | 39 | 6068 | |||
XM_017003890.1 | XP_016859379.1 | 39 | 6056 | |||
XM_017003891.1 | XP_016859380.1 | 39 | 6336 | |||
XM_017003892.1 | XP_016859381.1 | 27 | 5120 | |||
XM_017003893.1 | XP_016859382.1 | 40 | 6317 | |||
XM_017003894.1 | XP_016859383.1 | 18 | 2548 | |||
GenBank mRNA | AB058746.2 | - | 25 | 3522 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
