siRNA Details
Gene Symbol: UNC80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032504.1 | NP_115893.1 | 46 | 7094 | ||
NM_182587.3 | NP_872393.3 | 46 | 7079 | |||
XM_005246476.1 | XP_005246533.1 | 47 | 7289 | |||
XM_011511004.1 | XP_011509306.1 | 47 | 7334 | |||
XM_011511005.1 | XP_011509307.1 | 47 | 7331 | |||
XM_011511006.1 | XP_011509308.1 | 47 | 7328 | |||
XM_011511007.1 | XP_011509309.1 | 47 | 7298 | |||
XM_011511008.1 | XP_011509310.1 | 47 | 7334 | |||
XM_011511010.2 | XP_011509312.1 | 47 | 7334 | |||
XM_017003884.1 | XP_016859373.1 | 47 | 7319 | |||
XM_017003885.1 | XP_016859374.1 | 47 | 7292 | |||
XM_017003886.1 | XP_016859375.1 | 47 | 7334 | |||
XM_017003887.1 | XP_016859376.1 | 46 | 7130 | |||
XM_017003888.1 | XP_016859377.1 | 46 | 7088 | |||
XM_017003889.1 | XP_016859378.1 | 46 | 7085 | |||
XM_017003890.1 | XP_016859379.1 | 46 | 7073 | |||
XM_017003891.1 | XP_016859380.1 | 46 | 7353 | |||
XM_017003892.1 | XP_016859381.1 | 34 | 6137 | |||
XM_017003894.1 | XP_016859383.1 | 25 | 3565 | |||
GenBank mRNA | AB058746.2 | - | 32 | 4539 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
