siRNA Details
Gene Symbol: COL8A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001850.4 | NP_001841.2 | 4 | 573 | ||
NM_020351.3 | NP_065084.2 | 3 | 511 | |||
GenBank mRNA | AY550974.1 | - | 2 | 371 | ||
BC013581.1 | - | 4 | 551 | 3875911 | ||
BT009917.1 | - | 1 | 314 | |||
CA393523.1 | - | 3 | 497 | |||
CN484310.1 | - | 3 | 497 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
