siRNA Details
Gene Symbol: COL8A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001850.4 | NP_001841.2 | 5 | 2328 | ||
NM_020351.3 | NP_065084.2 | 4 | 2266 | |||
GenBank mRNA | AY550974.1 | - | 3 | 2129 | ||
BC013581.1 | - | 5 | 2306 | 3875911 | ||
BG938623.1 | - | 1 | 140 | |||
BT009917.1 | - | 2 | 2069 | |||
X57527.1 | - | 2 | 2069 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
