siRNA Details
Gene Symbol: TMEM44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011655.2 | NP_001011655.1 | 3 | 486 | ||
NM_001166305.1 | NP_001159777.1 | 3 | 486 | |||
NM_001166306.1 | NP_001159778.1 | 3 | 486 | |||
NM_138399.4 | NP_612408.3 | 3 | 486 | |||
XM_005269371.4 | XP_005269428.1 | 3 | 500 | |||
XM_011513318.2 | XP_011511620.1 | 3 | 389 | |||
XM_011513319.2 | XP_011511621.1 | 3 | 400 | |||
XM_011513320.2 | XP_011511622.1 | 3 | 510 | |||
XM_011513321.2 | XP_011511623.1 | 3 | 397 | |||
XM_011513322.2 | XP_011511624.1 | 3 | 507 | |||
XM_017007517.1 | XP_016863006.1 | 3 | 388 | |||
XM_017007518.1 | XP_016863007.1 | 3 | 383 | |||
GenBank mRNA | AK126914.1 | - | 3 | 225 | ||
AL547748.3 | - | 3 | 497 | |||
BC014883.1 | - | 3 | 346 | 3907191 | ||
BC111485.1 | - | 3 | 445 | 5767916 | ||
BC130297.1 | - | 3 | 390 | 40146328 | ||
BC144158.1 | - | 3 | 390 | 9052678 | ||
BC144159.1 | - | 3 | 390 | 9052679 | ||
BU521517.1 | - | 2 | 42 | |||
BU850042.1 | - | 3 | 449 | |||
BX419619.2 | - | 3 | 447 | |||
BX647418.1 | - | 3 | 245 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
