siRNA Details
Gene Symbol: CPLX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008220.1 | NP_001008221.1 | 4 | 1366 | ||
NM_006650.3 | NP_006641.1 | 5 | 1416 | |||
XM_005265798.1 | XP_005265855.1 | 4 | 1294 | |||
XM_005265799.1 | XP_005265856.1 | 4 | 1404 | |||
XM_011534419.1 | XP_011532721.1 | 4 | 1682 | |||
XM_017008964.1 | XP_016864453.1 | 4 | 1424 | |||
GenBank mRNA | AK124837.1 | - | 2 | 517 | ||
AK293819.1 | - | 5 | 1193 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
