siRNA Details
Gene Symbol: TRIM7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203293.2 | NP_976038.1 | 7 | 2175 | ||
NM_203294.1 | NP_976039.1 | 7 | 1789 | |||
NM_203295.1 | NP_976040.1 | 7 | 1857 | |||
NM_203296.1 | NP_976041.1 | 7 | 2066 | |||
NM_203297.1 | NP_976042.1 | 5 | 2223 | |||
XM_017009903.1 | XP_016865392.1 | 7 | 2214 | |||
GenBank mRNA | AF396651.1 | - | 7 | 2052 | ||
AF396652.1 | - | 7 | 1789 | |||
AF396653.1 | - | 7 | 1857 | |||
AF396654.1 | - | 7 | 1850 | |||
AF396655.1 | - | 5 | 2229 | |||
AK124617.1 | - | 2 | 3576 | |||
BC132863.1 | - | 7 | 2054 | 40146885 | ||
BC132867.1 | - | 7 | 2054 | 40146889 | ||
BM825056.1 | - | 1 | 322 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
