siRNA Details
Gene Symbol: TIAM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012454.3 | NP_036586.2 | 4 | 1632 | ||
GenBank mRNA | AK123859.1 | - | 5 | 1727 | ||
AK125459.1 | - | 1 | 91 | |||
BC137275.1 | - | 3 | 1505 | 9021272 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
