siRNA Details
Gene Symbol: ZC3HAV1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080660.3 | NP_542391.2 | 5 | 1140 | ||
GenBank mRNA | BC008842.2 | - | 5 | 1140 | 4136104 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
