siRNA Details
Gene Symbol: CXorf36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024689.2 | NP_078965.2 | 2 | 452 | ||
NM_176819.3 | NP_789789.2 | 2 | 452 | |||
XM_005272670.1 | XP_005272727.1 | 2 | 427 | |||
XM_006724559.1 | XP_006724622.1 | 2 | 427 | |||
GenBank mRNA | AK294472.1 | - | 1 | 21 | ||
AY358764.1 | - | 2 | 410 | |||
BP231945.1 | - | 2 | 403 | |||
DC332862.1 | - | 1 | 23 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
