siRNA Details
Gene Symbol: CXorf36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024689.2 | NP_078965.2 | 3 | 1256 | ||
GenBank mRNA | AK024165.1 | - | 3 | 1257 | ||
BC035375.1 | - | 3 | 1192 | |||
BC137528.1 | - | 3 | 1244 | |||
BC137529.1 | - | 3 | 1244 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
