siRNA Details
Gene Symbol: KRBOX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001129898.1 | NP_001123370.1 | 6 | 1875 | ||
NM_001129899.1 | NP_001123371.1 | 7 | 1919 | |||
NM_001129900.1 | NP_001123372.1 | 7 | 1904 | |||
NM_017776.2 | NP_060246.2 | 6 | 1860 | |||
GenBank mRNA | AK000351.1 | - | 6 | 1727 | ||
AK096413.1 | - | 6 | 1769 | |||
AK097159.1 | - | 1 | 2745 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
