siRNA Details
Gene Symbol: MAGEA12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166386.3 | NP_001159858.1 | 3 | 583 | ||
NM_001166387.3 | NP_001159859.1 | 3 | 491 | |||
NM_005367.6 | NP_005358.2 | 2 | 385 | |||
GenBank mRNA | AK314168.1 | - | 3 | 583 | ||
BC003408.2 | - | 2 | 373 | 3450217 | ||
BC098146.1 | - | 3 | 163 | |||
BQ436888.1 | - | 2 | 385 | |||
BT007108.1 | - | 1 | 229 | |||
CF123053.1 | - | 3 | 488 | |||
CR541775.1 | - | 1 | 229 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
