siRNA Details
Gene Symbol: MFSD14B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032558.2 | NP_115947.2 | 7 | 1018 | ||
XM_006717305.3 | XP_006717368.1 | 7 | 993 | |||
XM_011519114.2 | XP_011517416.1 | 7 | 709 | |||
XM_017015217.1 | XP_016870706.1 | 7 | 819 | |||
XM_017015218.1 | XP_016870707.1 | 7 | 746 | |||
XM_017015219.1 | XP_016870708.1 | 6 | 556 | |||
XM_017015220.1 | XP_016870709.1 | 6 | 570 | |||
XM_017015221.1 | XP_016870710.1 | 7 | 709 | |||
GenBank mRNA | AK027659.1 | - | 3 | 323 | ||
AK222836.1 | - | 3 | 333 | |||
AK300615.1 | - | 6 | 590 | |||
BC021093.1 | - | 2 | 128 | |||
BC106062.1 | - | 7 | 755 | 6162855 | ||
BX648846.1 | - | 4 | 432 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
