siRNA Details
Gene Symbol: EPG5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020964.2 | NP_066015.2 | 17, 18 | 3338 | ||
XM_011526122.1 | XP_011524424.1 | 17, 18 | 3338 | |||
XM_017025889.1 | XP_016881378.1 | 17, 18 | 3320 | |||
XM_017025890.1 | XP_016881379.1 | 17, 18 | 3320 | |||
XM_017025892.1 | XP_016881381.1 | 16, 17 | 3363 | |||
XM_017025893.1 | XP_016881382.1 | 9, 10 | 1210 | |||
GenBank mRNA | AB046852.1 | - | 17, 18 | 3273 | ||
AK023817.1 | - | 1, 2 | 17 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
