siRNA Details
Gene Symbol: EPG5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020964.2 | NP_066015.2 | 22 | 4036 | ||
XM_011526122.1 | XP_011524424.1 | 22 | 4036 | |||
XM_017025889.1 | XP_016881378.1 | 22 | 4018 | |||
XM_017025890.1 | XP_016881379.1 | 22 | 4018 | |||
XM_017025891.1 | XP_016881380.1 | 21 | 3895 | |||
XM_017025892.1 | XP_016881381.1 | 21 | 4061 | |||
XM_017025893.1 | XP_016881382.1 | 14 | 1908 | |||
GenBank mRNA | AB046852.1 | - | 22 | 3971 | ||
AK023817.1 | - | 6 | 715 | |||
BC130614.1 | - | 5 | 569 | |||
BC144104.1 | - | 5 | 569 | |||
BC144105.1 | - | 5 | 569 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001195633, AK028969, AK122536.
Gene Ontology Categories:
