siRNA Details
Gene Symbol: EPG5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020964.2 | NP_066015.2 | 20, 21 | 3791 | ||
XM_011526122.1 | XP_011524424.1 | 20, 21 | 3791 | |||
XM_017025889.1 | XP_016881378.1 | 20, 21 | 3773 | |||
XM_017025890.1 | XP_016881379.1 | 20, 21 | 3773 | |||
XM_017025891.1 | XP_016881380.1 | 19, 20 | 3650 | |||
XM_017025892.1 | XP_016881381.1 | 19, 20 | 3816 | |||
XM_017025893.1 | XP_016881382.1 | 12, 13 | 1663 | |||
GenBank mRNA | AB046852.1 | - | 20, 21 | 3726 | ||
AK023817.1 | - | 4, 5 | 470 | |||
BC130614.1 | - | 3, 4 | 324 | |||
BC144104.1 | - | 3, 4 | 324 | |||
BC144105.1 | - | 3, 4 | 324 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
