siRNA Details
Gene Symbol: ZNF547 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173631.3 | NP_775902.2 | 1 | 153 | ||
GenBank mRNA | AK055662.1 | - | 1 | 65 | ||
BC042681.1 | - | 1 | 22 | 4823926 |
Gene Symbol: TRAPPC2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002166.1 | 1 | 77 | |||
GenBank mRNA | AF058918.1 | - | 1 | 77 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
