siRNA Details
Gene Symbol: ZNF547 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173631.3 | NP_775902.2 | 4 | 849 | ||
GenBank mRNA | AK055662.1 | - | 4 | 761 | ||
AK291464.1 | - | 4 | 696 | |||
AL832545.1 | - | 4 | 717 | |||
BC042681.1 | - | 4 | 718 | 4823926 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
