siRNA Details
Gene Symbol: ZNF551 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270938.1 | NP_001257867.1 | 3 | 763 | ||
NM_138347.4 | NP_612356.2 | 3 | 885 | |||
NR_073102.1 | 2 | 761 | ||||
GenBank mRNA | AK091273.1 | - | 3 | 845 | ||
AK300465.1 | - | 2 | 723 | |||
AK307670.1 | - | 3 | 763 | |||
BC005868.2 | - | 3 | 817 | 3530863 | ||
BX538151.1 | - | 2 | 731 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
