siRNA Details
Gene Symbol: SPEF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024867.3 | NP_079143.3 | 29 | 4266 | ||
XM_005248376.3 | XP_005248433.1 | 29 | 4371 | |||
XM_005248377.3 | XP_005248434.1 | 29 | 4356 | |||
XM_011514135.2 | XP_011512437.1 | 29 | 4376 | |||
XM_011514136.2 | XP_011512438.1 | 29 | 4375 | |||
XM_011514137.2 | XP_011512439.1 | 29 | 4360 | |||
XM_011514138.2 | XP_011512440.1 | 29 | 4352 | |||
XM_011514139.2 | XP_011512441.1 | 28 | 4300 | |||
XM_011514140.2 | XP_011512442.1 | 28 | 4110 | |||
XM_011514141.2 | XP_011512443.1 | 30 | 4574 | |||
XM_011514142.1 | XP_011512444.1 | 29 | 4347 | |||
XM_017009880.1 | XP_016865369.1 | 28 | 4192 | |||
XM_017009881.1 | XP_016865370.1 | 29 | 5230 | |||
GenBank mRNA | AB051557.1 | - | 19 | 2607 | ||
AK027230.1 | - | 6 | 712 | |||
AK225586.1 | - | 6 | 712 | |||
AK225607.1 | - | 6 | 712 | |||
BX648691.1 | - | 2 | 107 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
