siRNA Details
Gene Symbol: SPEF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024867.3 | NP_079143.3 | 35 | 5319 | ||
XM_005248376.3 | XP_005248433.1 | 35 | 5424 | |||
XM_005248377.3 | XP_005248434.1 | 35 | 5409 | |||
XM_011514135.2 | XP_011512437.1 | 40 | 6251 | |||
XM_011514136.2 | XP_011512438.1 | 40 | 6250 | |||
XM_011514137.2 | XP_011512439.1 | 40 | 6235 | |||
XM_011514138.2 | XP_011512440.1 | 40 | 6227 | |||
XM_011514139.2 | XP_011512441.1 | 39 | 6175 | |||
XM_011514140.2 | XP_011512442.1 | 39 | 5985 | |||
XM_011514141.2 | XP_011512443.1 | 41 | 6449 | |||
XM_011514142.1 | XP_011512444.1 | 35 | 5400 | |||
XM_017009880.1 | XP_016865369.1 | 39 | 6067 | |||
XM_017009881.1 | XP_016865370.1 | 40 | 7105 | |||
GenBank mRNA | AB051557.1 | - | 25 | 3660 | ||
AK026817.1 | - | 11 | 1823 | |||
AK027230.1 | - | 12 | 1765 | |||
AK225586.1 | - | 12 | 1765 | |||
AK225607.1 | - | 12 | 1765 | |||
BX648691.1 | - | 13 | 2827 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
