siRNA Details
Gene Symbol: SYCP2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040274.2 | NP_001035364.2 | 26, 27 | 2551 | ||
GenBank mRNA | AK128130.1 | - | 26, 27 | 2605 | ||
AM293674.1 | - | 26, 27 | 2270 | |||
BC012225.1 | - | 16, 17 | 1448 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
