siRNA Details
Gene Symbol: CCDC144CP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_023380.1 | 14 | 3884 | |||
GenBank mRNA | AK295707.1 | - | 2 | 197 | ||
DA321951.1 | - | 2 | 197 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
