siRNA Details
Gene Symbol: TRMT44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152544.2 | NP_689757.2 | 6 | 1164 | ||
XM_005247950.3 | XP_005248007.1 | 6 | 1161 | |||
XM_011513406.2 | XP_011511708.1 | 6 | 1193 | |||
XM_011513407.2 | XP_011511709.1 | 6 | 1193 | |||
XM_011513408.1 | XP_011511710.1 | 7 | 1305 | |||
XM_011513409.2 | XP_011511711.1 | 7 | 1276 | |||
XM_011513410.2 | XP_011511712.1 | 6 | 1195 | |||
XM_017007811.1 | XP_016863300.1 | 6 | 1194 | |||
GenBank mRNA | AK093044.1 | - | 2 | 170 | ||
BC035655.1 | - | 6 | 664 | 5588410 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
