siRNA Details
Gene Symbol: ANKRD26P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026556.1 | 13 | 2991 | |||
GenBank mRNA | AK125968.1 | - | 1 | 86 | ||
BC070117.1 | - | 13 | 2991 | |||
BX648636.1 | - | 3 | 774 | |||
CR749347.1 | - | 6 | 1614 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
