siRNA Details
Gene Symbol: TARP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003799.1 | NP_001003799.1 | 2 | 220 | ||
NM_001003806.1 | NP_001003806.1 | 2 | 220 | |||
XM_011515407.2 | XP_011513709.1 | 3 | 440 | |||
XM_017012226.1 | XP_016867715.1 | 3 | 375 | |||
XM_017012227.1 | XP_016867716.1 | 2 | 430 | |||
XM_017012228.1 | XP_016867717.1 | 4 | 821 | |||
XM_017012229.1 | XP_016867718.1 | 2 | 642 | |||
XM_017012230.1 | XP_016867719.1 | 2 | 246 | |||
GenBank mRNA | AF151103.1 | - | 2 | 220 | ||
BC030554.1 | - | 4 | 904 | |||
BC105589.1 | - | 3 | 438 | 4272944 | ||
BF674593.1 | - | 2 | 210 | |||
JQ956403.1 | - | 3 | 362 | |||
KC170727.1 | - | 5 | 577 | |||
M16804.1 | - | 3 | 499 | |||
M17324.1 | - | 1 | 159 | |||
M17325.1 | - | 1 | 139 | |||
M27331.1 | - | 1 | 160 | |||
M27332.1 | - | 1 | 160 | |||
M27334.1 | - | 1 | 160 | |||
M30894.1 | - | 4 | 699 | |||
S73842.1 | - | 3 | 400 | |||
X06774.1 | - | 4 | 680 | |||
X06775.1 | - | 2 | 250 | |||
X06776.1 | - | 3 | 435 | |||
X72500.1 | - | 4 | 586 | |||
X74774.1 | - | 3 | 686 | |||
Y00790.1 | - | 4 | 598 |
Gene Symbol: TRGC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | M13231.1 | - | 4 | 569 | ||
M17323.1 | - | 1 | 159 |
Gene Symbol: TRGV9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC062761.1 | - | 2 | 215 | ||
BC072387.1 | - | 3 | 537 | |||
BC072396.1 | - | 5 | 963 | |||
M16768.1 | - | 4 | 723 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
