siRNA Details
Gene Symbol: TARP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017012227.1 | XP_016867716.1 | 1 | 226 | ||
XM_017012229.1 | XP_016867718.1 | 1 | 438 | |||
GenBank mRNA | BC030554.1 | - | 3 | 700 | ||
S60779.1 | - | 2 | 496 | |||
S60780.1 | - | 2 | 482 | |||
X74774.1 | - | 2 | 482 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
