siRNA Details
Gene Symbol: TSGA13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304968.1 | NP_001291897.1 | 2 | 628 | ||
NM_052933.3 | NP_443165.1 | 1 | 51 | |||
GenBank mRNA | AF431815.1 | - | 1 | 51 | ||
AK093329.1 | - | 2 | 627 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
