siRNA Details
Gene Symbol: TSGA13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304968.1 | NP_001291897.1 | 4 | 938 | ||
NM_052933.3 | NP_443165.1 | 3 | 543 | |||
GenBank mRNA | AF294347.1 | - | 3 | 261 | ||
AF431815.1 | - | 3 | 543 | |||
AK093329.1 | - | 4 | 937 | |||
BC038415.1 | - | 3 | 543 | 5172129 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
