siRNA Details
Gene Symbol: F11R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016946.4 | NP_058642.1 | 1 | 296 | ||
GenBank mRNA | AF154005.1 | - | 1 | 88 | ||
AF172398.2 | - | 1 | 172 | |||
AF207907.1 | - | 1 | 41 | |||
AK026665.1 | - | 1 | 87 | |||
AK075152.1 | - | 1 | 96 | |||
AK304412.1 | - | 1 | 49 | |||
AL136649.1 | - | 1 | 100 | |||
AY358896.1 | - | 1 | 77 | |||
BC001533.2 | - | 1 | 70 | 2990019 | ||
BT020103.1 | - | 1 | 26 | |||
CR533512.1 | - | 1 | 26 | |||
DA907460.1 | - | 1 | 296 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
