siRNA Details
Gene Symbol: F11R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016946.4 | NP_058642.1 | 5 | 829 | ||
GenBank mRNA | AF154005.1 | - | 5 | 501 | ||
AF172398.2 | - | 5 | 705 | |||
AF207907.1 | - | 5 | 574 | |||
AK026665.1 | - | 5 | 620 | |||
AK075152.1 | - | 5 | 629 | |||
AK299487.1 | - | 5 | 610 | |||
AK304412.1 | - | 4 | 435 | |||
AL136649.1 | - | 5 | 633 | |||
AY358896.1 | - | 5 | 610 | |||
BC001533.2 | - | 5 | 603 | 2990019 | ||
BT020103.1 | - | 5 | 559 | |||
CR533512.1 | - | 5 | 559 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
