siRNA Details
Gene Symbol: ZNF155 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001260486.1 | NP_001247415.1 | 5 | 1333 | ||
NM_001260487.1 | NP_001247416.1 | 5 | 1216 | |||
NM_001260488.1 | NP_001247417.1 | 6 | 1319 | |||
NM_003445.3 | NP_003436.3 | 5 | 1307 | |||
NM_198089.2 | NP_932355.2 | 5 | 1230 | |||
XM_005259215.3 | XP_005259272.1 | 5 | 1377 | |||
XM_011527278.2 | XP_011525580.1 | 6 | 1262 | |||
XM_011527279.2 | XP_011525581.1 | 6 | 2322 | |||
XM_017027248.1 | XP_016882737.1 | 6 | 1354 | |||
GenBank mRNA | AF187986.1 | - | 5 | 1260 | ||
AK022875.1 | - | 5 | 1197 | |||
AK297360.1 | - | 6 | 1295 | |||
AK297384.1 | - | 4 | 1043 | |||
AK314510.1 | - | 5 | 1212 | |||
BC075050.2 | - | 4 | 1101 | 30915579 | ||
BC075051.2 | - | 4 | 1101 | 30915363 | ||
BC126377.1 | - | 4 | 1155 | 8992093 | ||
BC130381.1 | - | 4 | 1155 | 40146412 | ||
U09852.1 | - | 1 | 95 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
