siRNA Details
Gene Symbol: C21orf58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286463.1 | NP_001273392.1 | 7 | 1578 | ||
NM_001286476.1 | NP_001273405.1 | 9 | 2557 | |||
NM_001286477.1 | NP_001273406.1 | 8 | 2219 | |||
NM_058180.4 | NP_478060.2 | 8 | 2591 | |||
XM_005261148.4 | XP_005261205.1 | 9 | 2294 | |||
XM_011529621.2 | XP_011527923.1 | 8 | 2574 | |||
XM_011529623.2 | XP_011527925.1 | 8 | 4579 | |||
XM_011529624.2 | XP_011527926.1 | 8 | 1454 | |||
XM_011529625.2 | XP_011527927.1 | 8 | 1728 | |||
XM_011529626.2 | XP_011527928.1 | 9 | 4925 | |||
XM_011529627.2 | XP_011527929.1 | 9 | 5005 | |||
XM_017028388.1 | XP_016883877.1 | 9 | 4756 | |||
XM_017028389.1 | XP_016883878.1 | 10 | 4642 | |||
XM_017028390.1 | XP_016883879.1 | 9 | 2923 | |||
XM_017028391.1 | XP_016883880.1 | 8 | 2746 | |||
XM_017028392.1 | XP_016883881.1 | 10 | 1943 | |||
XM_017028393.1 | XP_016883882.1 | 9 | 1742 | |||
GenBank mRNA | AK056387.1 | - | 8 | 2564 | ||
AK096641.1 | - | 7 | 2495 | |||
AK098098.1 | - | 7 | 1515 | |||
BC026042.1 | - | 9 | 2532 | |||
BM475055.1 | - | 1 | 331 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
