siRNA Details
Gene Symbol: FAM189B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267608.1 | NP_001254537.1 | 1 | 587 | ||
NM_006589.2 | NP_006580.2 | 1 | 587 | |||
NM_198264.1 | NP_937995.1 | 1 | 587 | |||
XM_005244845.1 | XP_005244902.1 | 1 | 581 | |||
GenBank mRNA | BC006493.2 | - | 1 | 67 | 2966515 | |
BC007444.2 | - | 1 | 67 | 2966515 | ||
BC008854.2 | - | 1 | 452 | |||
BC010471.1 | - | 1 | 491 | 4179576 | ||
BC017262.2 | - | 1 | 295 | |||
BC040136.1 | - | 1 | 64 | 6061151 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
