siRNA Details
Gene Symbol: FAM189B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267608.1 | NP_001254537.1 | 5 | 1333 | ||
NM_006589.2 | NP_006580.2 | 6 | 1387 | |||
NM_198264.1 | NP_937995.1 | 3 | 1099 | |||
XM_005244845.1 | XP_005244902.1 | 6 | 1384 | |||
GenBank mRNA | AL353941.1 | - | 1 | 63 | ||
BC006493.2 | - | 6 | 867 | 2966515 | ||
BC007444.2 | - | 6 | 867 | 2966515 | ||
BC008854.2 | - | 3 | 964 | |||
BC010471.1 | - | 6 | 1291 | 4179576 | ||
BC040136.1 | - | 3 | 576 | 6061151 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
