siRNA Details
Gene Symbol: FAM189B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267608.1 | NP_001254537.1 | 10 | 2326 | ||
NM_006589.2 | NP_006580.2 | 11 | 2380 | |||
NM_198264.1 | NP_937995.1 | 8 | 2092 | |||
XM_005244845.1 | XP_005244902.1 | 11 | 2377 | |||
GenBank mRNA | AF070550.1 | - | 5 | 775 | ||
AL353941.1 | - | 6 | 1164 | |||
BC006297.1 | - | 3 | 792 | |||
BC006493.2 | - | 11 | 1860 | 2966515 | ||
BC007444.2 | - | 11 | 1860 | 2966515 | ||
BC010471.1 | - | 11 | 2284 | 4179576 | ||
BC017262.2 | - | 4 | 911 | |||
BC040136.1 | - | 8 | 1569 | 6061151 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
