siRNA Details
Gene Symbol: MUM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032853.3 | NP_116242.2 | 14 | 2361 | ||
NR_024247.1 | 14 | 2430 | ||||
XM_011528380.2 | XP_011526682.1 | 15 | 2453 | |||
XM_011528385.2 | XP_011526687.1 | 15 | 2552 | |||
GenBank mRNA | AF151045.1 | - | 3 | 296 | ||
AK075241.1 | - | 8 | 991 | |||
AL360266.1 | - | 2 | 241 | |||
BC008098.1 | - | 6 | 845 | |||
BC082987.1 | - | 5 | 812 | |||
BC110874.1 | - | 13 | 2327 | 4933735 | ||
BC130443.1 | - | 14 | 2390 | 40146474 | ||
BC144138.1 | - | 14 | 2434 | 9052658 | ||
BC144147.1 | - | 14 | 2390 | |||
BG723598.1 | - | 2 | 201 | |||
CR457395.1 | - | 1 | 58 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
