siRNA Details
Gene Symbol: TNFSF13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198622.1 | NP_001185551.1 | 5 | 1919 | ||
NM_001198623.1 | NP_001185552.1 | 5 | 1916 | |||
NM_001198624.1 | NP_001185553.1 | 6 | 1224 | |||
NM_003808.3 | NP_003799.1 | 6 | 2000 | |||
NM_172087.2 | NP_742084.1 | 5 | 1952 | |||
NM_172088.2 | NP_742085.1 | 7 | 1819 | |||
NR_073490.2 | 4 | 1873 | ||||
GenBank mRNA | AF114012.1 | - | 5 | 1401 | ||
AF114013.1 | - | 7 | 1324 | |||
AK090698.1 | - | 5 | 1768 | |||
AK225556.1 | - | 6 | 1544 | |||
AY081050.1 | - | 6 | 1524 | |||
BC008042.2 | - | 7 | 1174 | 3837834 |
Gene Symbol: TNFSF12-TNFSF13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172089.3 | NP_742086.1 | 11 | 1588 | ||
GenBank mRNA | AY081051.1 | - | 11 | 1547 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
