siRNA Details
Gene Symbol: ZNF326 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320185.1 | NP_001307114.1 | 2, 3 | 235 | ||
NM_181781.3 | NP_861446.2 | 2, 3 | 235 | |||
NM_182975.3 | NP_892020.1 | 2, 3 | 235 | |||
NM_182976.3 | NP_892021.1 | 2, 3 | 235 | |||
XM_005270779.4 | XP_005270836.1 | 2, 3 | 215 | |||
XM_011541290.2 | XP_011539592.1 | 2, 3 | 230 | |||
GenBank mRNA | AK000410.1 | - | 2, 3 | 202 | ||
AK297073.1 | - | 2, 3 | 160 | |||
AK303707.1 | - | 2, 3 | 162 | |||
BC014899.1 | - | 2, 3 | 193 | |||
BC052645.1 | - | 2, 3 | 209 | |||
BC053888.1 | - | 2, 3 | 196 | |||
BC070341.1 | - | 2, 3 | 179 | |||
BC090866.1 | - | 2, 3 | 229 | 6503526 | ||
CN299618.1 | - | 2, 3 | 245 | |||
DB111509.1 | - | 2, 3 | 211 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
