siRNA Details
Gene Symbol: ZNF326 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320185.1 | NP_001307114.1 | 8 | 957 | ||
NM_181781.3 | NP_861446.2 | 6 | 760 | |||
NM_182976.3 | NP_892021.1 | 8 | 1224 | |||
XM_005270779.4 | XP_005270836.1 | 7 | 798 | |||
XM_011541290.2 | XP_011539592.1 | 8 | 930 | |||
GenBank mRNA | AK297073.1 | - | 8 | 1149 | ||
AK303707.1 | - | 6 | 687 | |||
BC014899.1 | - | 8 | 915 | |||
BC070341.1 | - | 8 | 1168 | |||
BC090866.1 | - | 8 | 1218 | 6503526 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
