siRNA Details
Gene Symbol: ZNF326 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320185.1 | NP_001307114.1 | 12 | 1868 | ||
NM_181781.3 | NP_861446.2 | 10 | 1671 | |||
NM_182976.3 | NP_892021.1 | 12 | 2135 | |||
XM_005270779.4 | XP_005270836.1 | 11 | 1709 | |||
XM_011541290.2 | XP_011539592.1 | 12 | 1841 | |||
GenBank mRNA | BC090866.1 | - | 12 | 2129 | 6503526 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
