siRNA Details
Gene Symbol: DENND1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195215.1 | NP_001182144.1 | 12, 13 | 1161 | ||
NM_001300858.1 | NP_001287787.1 | 12, 13 | 1312 | |||
NM_144977.4 | NP_659414.2 | 12, 13 | 1161 | |||
NR_125340.1 | 11, 12 | 1101 | ||||
XM_005244931.2 | XP_005244988.1 | 11, 12 | 1101 | |||
XM_006711192.2 | XP_006711255.1 | 13, 14 | 1071 | |||
XM_006711193.3 | XP_006711256.1 | 13, 14 | 1203 | |||
XM_011509246.2 | XP_011507548.1 | 12, 13 | 1022 | |||
XM_011509247.1 | XP_011507549.1 | 11, 12 | 962 | |||
XM_011509248.2 | XP_011507550.1 | 12, 13 | 955 | |||
XM_011509249.2 | XP_011507551.1 | 13, 14 | 1260 | |||
XM_011509251.2 | XP_011507553.1 | 12, 13 | 1024 | |||
XM_017000470.1 | XP_016855959.1 | 13, 14 | 1149 | |||
XM_017000471.1 | XP_016855960.1 | 12, 13 | 1011 | |||
GenBank mRNA | AK091207.1 | - | 11, 12 | 915 | ||
AL831839.1 | - | 6, 7 | 448 | |||
BC022561.1 | - | 12, 13 | 1014 | 4793412 | ||
BC063877.1 | - | 12, 13 | 993 | 30377843 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
